Orbital Shaker, KS 501 digital, IKA® WORKS
2526401
33994-212EA 202681.69 MXN
33994-212
Orbital Shaker, KS 501 digital, IKA® WORKS
Sleek, quiet shaker with a large base designed for loads up to 15kg (33 lbs.), including attachment
Speed is digitally displayed. Continuous operation or timed shaking up to 56 minutes.
Información para pedidos: Platforms sold separately. The stainless steel fixing clip support 33994-266 measures 47.6Wx46Lx9.5H cm (183/4x181/8x33/4") and holds up to 50 flasks; for fixing clips, see 33994-246. The dish attachment 33994-268 measures 45Wx45Lx4.4H cm (173/4x173/4x13/4") and accommodates dishes or flat bottom vessels.