Filter Funnel Manifolds, Cytiva (Formerly Pall Lab)
15402 15403 15403 15402
28145-348EA 58561.61 MXN
CA28145-349 CA28145-348 28145-348 28145-349
Filter Funnel Manifolds, Cytiva (Formerly Pall Lab)
For use in vacuum filtration of liquids for analysis of microbiological or particulate contamination, and scintillation studies.
  • For vacuum filtration
  • On/off valve
  • Aluminum manifold
Can accommodate 25 or 47mm filter funnels. On/off control valves allow independent operation of each station. Anodized aluminum manifold body construction is especially suited for applications where chemical compatibility is critical and easy clean-up is desired. Chrome-plated brass valves. Viton® rubber valve O-rings and 6.4mm (1/4") nylon adapter. Stainless steel, 6.4mm (1/4") MNPT drain plugs.