Nylaflo® Membrane Disc Filters
Proveedor: Cytiva
Nylaflo®
66607 66602 66607 66606 66608 66609 66601 66604 66608 66604 66610 66602 66610 66603 66603 66601 66609
28140-040EA 183.8 MXN
28140-040 28140-041 CA28140-044 CA28140-143 CA28140-141 CA28140-042 28140-143 28140-044 CA28140-120 28140-042 28140-141 CA28140-040 28140-120 CA28140-041 CA28140-028 28140-108 28140-028
Nylaflo® Membrane Disc Filters
Filtros Filtros de membranas
Useful for a wide range of both aqueous and solvent-based applications.
  • Hydrophilic
  • Excellent chemical compatibility
  • Naturally wettable
Hydrophilic nylon filters offer excellent chemical compatibility with esters, bases, and alcohols
Naturally wettable to aqueous solutions. HPLC certified for low levels of UV-absorbing extractables.
Not recommended for acids >1N or halogenated solvents.