Nalgene® Stopcocks, Polypropylene with Teflon® Resin TFE Plug, Thermo Scientific
Proveedor: Thermo Scientific
Nalgene®
6460-0004 6460-0002
59096-055CS 13355.5 MXN
59096-055 59096-000
Nalgene® Stopcocks, Polypropylene with Teflon® Resin TFE Plug, Thermo Scientific
Válvulas de cierre
Serrated tubulation on each end
Accepts 6.4–7.9mm (1/45/16") tubing. Autoclavable.