PTFE Stopcock Plugs, Straight Bore, Kimble Chase
Proveedor: DWK Life Sciences
41500F-6 41500F-2 41500F-4
89002-170CS 24619.42 MXN
89002-172 59518-382 89002-170
PTFE Stopcock Plugs, Straight Bore, Kimble Chase
Válvulas de cierre
For separatory funnels and funnel-fill burets requiring [PS] PTFE stopcock plugs
Complete with nut and washers (1 PTFE, 1 rubber).