High Vacuum Stopcock, Easy-Action Plug, 90 Degree, Ace Glass
Proveedor: Ace Glass
8195-44 8195-32 8195-46 8195-33 8195-40 8195-246 8195-240 8195-238 8195-31 8195-34 8195-21 8195-48 8195-236 8195-12 8195-47 8195-242 8195-23 8195-45 8195-63 8195-36 8195-244 8195-41 8195-04 8195-30 8195-24 8195-43 8195-61 8195-09
89062-594EA 2667.86 MXN
76107-038 89062-592 89062-594 89062-630 89062-596 89062-610 89062-612 89062-634 89062-414 89062-614 89062-636 89062-616 89062-638 89062-618 89062-584 76107-030 76107-032 89062-422 76107-034 89062-600 89062-622 76107-036 89062-602 89062-626 89062-406 89062-604 89062-606 89062-408
High Vacuum Stopcock, Easy-Action Plug, 90 Degree, Ace Glass
Válvulas de cierre
These are high vacuum stopcocks with arms at 90 degrees with variable openings from 0-3 mm to 0-15 mm
An ultimate vacuum of 10-7 can be realized with the standard three O-rings. Smooth acting semi-needle valve permits fine adjustment of opening. Plugs available in bakeable PTFE or glass with Nylon handle. Reference marks on body aid in repetitive setting. Supplied with FETFE O-rings. Plug only and replacement O-rings available