Nalgene® Cylindrical Tanks with Spigot, LLDPE, Thermo Scientific
Proveedor: Thermo Scientific
Nalgene®
54102-0007 54102-0015 54102-0005 54102-0055 54102-0030 54102-0010
60454-034EA 11388.96 MXN
60454-034 60454-031 60454-040 60454-083 60454-061 60454-028
Nalgene® Cylindrical Tanks with Spigot, LLDPE, Thermo Scientific
Depósitos Depósitos cilíndricos
Tanks feature polypropylene needle spigot, stepped flange at rim, and overlapping skirted cover
Graduated in gallons and liters, with the exception of tanks 60454-031 and 60454-034, which are graduated in gallons only. Good chemical resistance and tolerant of a wide range of temperatures. Not autoclavable.