HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Proveedor: Air-Tite
HENKE-JECT® NORM-JECT®
4200.000V0 4850001000 NJ-4606205-02 NJ-4606067-02 NJ-4606027-02 4830001000 4100.000V0 NJ-9166017-02 NJ-4606108-02 4010.200V0 4050.000VZ 4020.000V0
53548-008CS 9047.14 MXN
53548-008 76459-478 53548-006 53548-005 53548-003 53548-024 76459-480 53548-001 53548-010 76459-482 76459-474 76459-476
HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Jeringas Jeringas médicas
NORM-JECT® syringes feature medical-grade plastics, including a polypropylene barrel and a polyethylene plunger. Great value at reasonable costs.
  • Sterile
  • Certified DNA free
  • Latex free
  • Rubber free
  • Silicone free
  • Styrene free
  • DEHP free
These syringes are the choice for any situation needing an inert, non-reactive syringe. Because of their composition, these syringes are indicated for Nuclear Medicine, Amniocentisis, IVF, embryo-transfer, chromatography and many laboratory procedures. They are more chemically resistant than rubber tipped syringes. These inert syringes do not contain contaminates often associated with disposables.
The syringes can be used for sampling, mixing, and filtering. They are smooth drawing with a positive safety stop to prevent accidental spills. They feature accurate, easy-to-read increments. Their dose saver design minimizes waste, saving up to 4.5 ml of medication per 100 syringes used, as compared to non-low-deadspace Luer syringes (Actual savings may vary).
Certificaciones: Meets ISO Standards, CE registered, FDA registered.
Envase: Sterile, individually wrapped.