Perfektum® Hypodermic Matched Numbered Glass Syringes, Cadence Science®
Proveedor: Cadence Science
Perfectum®
5127 5137 5101
20068-074EA 1081.24 MXN
20068-074 20068-076 20068-068
Perfektum® Hypodermic Matched Numbered Glass Syringes, Cadence Science®
Jeringas Jeringas médicas
Matched numbered syringes feature frosted individually lapped barrels and plungers which can withstand higher temperatures
These syringes are individually boxed and have matching serial numbers on mating barrels and plungers.
Información para pedidos: For a collection of related large volume Metal Luer Lock Tip syringes, see the Perfectum® Jumbo Syringes series.