Tissue-Tek® Slide Staining Set, Sakura
Proveedor: Sakura
Tissue-Tek®
4451
25608-902EA 17415.65 MXN
25608-902
Tissue-Tek® Slide Staining Set, Sakura
Cajas de tinción
Designed for histological or cytological staining
Provides simultaneous use of up to 12 solution wells, each deep enough to permit total immersion of 2.5x7.6cm (1x3") slides. Snap-locking cover on solvent-resistant metal frame prevents spills.
Información para pedidos: Complete set includes 10 white polypropylene dishes, 2 green solvent-resistant dishes, 12 lids, 12-unit metal carrier, and drain tray/cover. Additional staining dishes and 24-slide holder with detachable handle are sold separately.