Perfectum® Jumbo Syringes, Cadence Science
5177 5333 5331
20068-528EA 8759.62 MXN
20068-526 20068-528 20068-530
Perfectum® Jumbo Syringes, Cadence Science
These precision-made syringes are ideal for efficiently dispensing larger volumes of corrosive, toxic, and noxious liquids in aspirating, infusing, pipetting, sampling and other critical procedures
Lock tip syringes feature large chrome-plated metal Luer-lock tips. The 250mL jumbo syringe is specifically used for air sample work, infusion and pumps for liquid metering applications.
They are suitable for biomedical, environmental, laboratory, research, and industrial applications. Syringes are chemical-resistant. They are made of fully annealed glass that is resistant to thermal shock.