SP Wilmad-LabGlass Pennyhead [ST] Stoppers, Flask Length, SP Industries
Proveedor: SP Industries
LG-10320-114 LG-10320-110 LG-10320-112 LG-10320-108 LG-10320-104 LG-10320-100 LG-10320-106 LG-10320-102
14212-466EA 332.36 MXN
14212-540 14212-466 14212-344 14211-190 14212-522 14212-426 14210-838 14211-308
SP Wilmad-LabGlass Pennyhead [ST] Stoppers, Flask Length, SP Industries
Tapones Tapones de Vidrio
Glass flask length stoppers may be used with volumetric flasks and separatory funnels
Sizes 9 through 27 are solid. Sizes 32 and 38 are hollow.