VWR® Advanced Hot Plates
Proveedor: VWR
VWR®
984VW4AHPUSA 984VW0AHPUSA 984VW4CHPUSA 984VW0CHPUSA 984VW7AHPUSA 984VW7CHPUSA
97042-622EA 11189.65 MXN
97042-578 97042-622 97042-658 97042-582 97042-618 97042-662
VWR® Advanced Hot Plates
Calentadores Placas calefactoras
Advanced hot plates are designed to deliver accurate and repeatable results in general-purpose laboratory applications.
  • Excellent temperature uniformity
  • Industry Leading Safety Features
  • Cool touch, chemical-resistant housing
  • LED display for temperature
  • Now with Best-in-Class, Exclusive 5-Year Warranty
These microprocessor controlled units feature a front-facing control panel with an Easy-to-read LED that displays set-point temperature
Hot plates are available with ceramic or aluminum top plate material. Reflective white ceramic tops are chemical-resistant and easy to clean. Durable aluminum tops will not crack or chip, and offer a more even heating surface. A built-in memory recall provides users with the last set temperature, even after the unit has been turned off. For additional safety, a hot symbol warning light is illuminated when the hot plate is in use, and remains lit until the top plate is sufficiently cooled.Units are equipped with redundant safety controls, which will shut down unit in case of heater malfunction.
Enhanced electronics regulate the robust heater, which brings samples to the desired temperature quickly and efficiently. The low-profile design utilizes minimal bench space and permits use within a fume hood. Unit housing is constructed of a heat- and chemical-resistant polymer that remains cool to the touch during operation. The spill-resistant design channels fluids away from internal components in the event of spills.
Certificaciones: RoHS certified. TUV listed. CE marked.
Información para pedidos: Units are supplied with a 234 cm (92") three-wire cord and plug. The optional support rod and clamp kit includes a 45.7 cm (18") long stainless steel support rod, a thermometer/temperature probe extension clamp, a three-prong dual-adjust swivel clamp, and hook connector. 230V, 50/60Hz models are also available; contact your Avantor sales representative for more information.