Synthware Syringe Needles, Stainless Steel, Kemtech America
Proveedor: Kemtech America
Synthware
S389200 S386200 S386150 S380230 S380216 S389300 S389150 S380221 S386300
60004-796EA 277.99 MXN
60004-792 60004-794 76264-420 76264-134 76264-138 76264-136 76264-422 60004-796 76264-418
Synthware Syringe Needles, Stainless Steel, Kemtech America
Agujas de jeringas
Stainless steel needles are designed with extra length for solvent retrieval.
  • Stainless steel construction
  • Luer lock design
These long needles are ideal for solvent retrieval.