Order Entry
VWR® Ultrasonic Cleaners
undefined
VWR® Ultrasonic Cleaners
  • Catalog No:
  • 97043-948
  • 97043-940
  • 97043-984
  • 97043-964
  • 97044-010
  • 97043-944
  • 97043-988
  • 97043-968
  • 97043-932
  • 97043-976
  • 97043-956
  • 97044-002
  • 97043-936
  • 97043-980
  • 97043-960
  • 97044-014
  • 97043-992
  • 97043-972
  • 97043-952
  • Frequency:
  • 35 kHz
  • 35 kHz
  • 35 kHz
  • 35 kHz
  • 35 kHz
  • 35 kHz
  • 35 kHz
  • 35 kHz
  • 35 kHz
  • 35 kHz
  • 35 kHz
  • 35 kHz
  • 35 kHz
  • 35 kHz
  • 35 kHz
  • 35 kHz
  • 35 kHz
  • 35 kHz
  • 35 kHz
  • Temperature range:
  • 20 to 69°C (68 to 156°F)
  • 20 to 69°C (68 to 156°F)
  • 20 to 69°C (68 to 156°F)
  • 20 to 69°C (68 to 156°F)
  • 20 to 69°C (68 to 156°F)
  • 20 to 69°C (68 to 156°F)
  • 20 to 69°C (68 to 156°F)
  • 20 to 69°C (68 to 156°F)
  • 20 to 69°C (68 to 156°F)
  • 20 to 69°C (68 to 156°F)
  • 20 to 69°C (68 to 156°F)
  • 20 to 69°C (68 to 156°F)
  • 20 to 69°C (68 to 156°F)
  • 20 to 69°C (68 to 156°F)
  • 20 to 69°C (68 to 156°F)
  • 20 to 69°C (68 to 156°F)
  • 20 to 69°C (68 to 156°F)
  • 20 to 69°C (68 to 156°F)
  • 20 to 69°C (68 to 156°F)
  • Timer:
  • 1 - 99 minutes
  • 1 - 99 minutes
  • 1 - 99 minutes
  • 1 - 99 minutes
  • 1 - 99 minutes
  • 1 - 99 minutes
  • 1 - 99 minutes
  • 1 - 99 minutes
  • 1 - 99 minutes
  • 1 - 99 minutes
  • 1 - 99 minutes
  • 1 - 99 minutes
  • 1 - 99 minutes
  • 1 - 99 minutes
  • 1 - 99 minutes
  • 1 - 99 minutes
  • 1 - 99 minutes
  • 1 - 99 minutes
  • 1 - 99 minutes
Ultrasonic Cleaner with Digital Timer
  •  
    Capacity
    Electrical
    Tank Dimensions
  • Ultrasonic Cleaner with Digital Timer
    Ultrasonic Cleaner with Digital Timer
    97043-964
    2.8 L (0.7 gal.)
    117 V, 60 Hz
    L×W×D: 24×14×10 cm (9⁷⁄₁₆×5½×3¹⁵⁄₁₆")
    Each (1UN)
    Each (1UN) - $10,926.64
    $10,926.64
    Each (1UN)
       $10,926.64
    cUL Listed
    UL Listed
    Accessories
    Specifications
    Environmentally Preferable
    Frequency
    35 kHz
    Temperature range
    20 to 69°C (68 to 156°F)
    Timer
    1 - 99 minutes
    cUL Listed
    UL Listed
  • Ultrasonic Cleaner with Digital Timer
    Ultrasonic Cleaner with Digital Timer
    97043-968
    5.7 L (1.5 gal.)
    117 V, 60 Hz
    L×W×D: 30×15×15 cm (11¹³⁄₁₆×5²⁹⁄₃₂×5²⁹⁄₃₂")
    Each (1UN)
    Each (1UN) - $15,830.33
    $15,830.33
    Each (1UN)
       $15,830.33
    cUL Listed
    UL Listed
    Accessories
    Specifications
    Environmentally Preferable
    Frequency
    35 kHz
    Temperature range
    20 to 69°C (68 to 156°F)
    Timer
    1 - 99 minutes
    cUL Listed
    UL Listed
  • Ultrasonic Cleaner with Digital Timer
    Ultrasonic Cleaner with Digital Timer
    97043-976
    13.8 L (3.6 gal.)
    117 V, 60 Hz
    L×W×D: 33×30×15 cm (13×11¹³⁄₁₆×5²⁹⁄₃₂")
    Each (1UN)
    Each (1UN) - $28,196.08
    $28,196.08
    Each (1UN)
       $28,196.08
    cUL Listed
    UL Listed
    Accessories
    Specifications
    Environmentally Preferable
    Frequency
    35 kHz
    Temperature range
    20 to 69°C (68 to 156°F)
    Timer
    1 - 99 minutes
    cUL Listed
    UL Listed
  • Ultrasonic Cleaner with Digital Timer
    Ultrasonic Cleaner with Digital Timer
    97043-984
    28.4 L (7.5 gal.)
    117 V, 60 Hz
    L×W×D: 50×30×20 cm (19¹¹⁄₁₆×11¹³⁄₁₆×7⅞")
    Each (1UN)
    Each (1UN) - $42,820.91
    $42,820.91
    Each (1UN)
       $42,820.91
    cUL Listed
    UL Listed
    Accessories
    Specifications
    Environmentally Preferable
    Frequency
    35 kHz
    Temperature range
    20 to 69°C (68 to 156°F)
    Timer
    1 - 99 minutes
    cUL Listed
    UL Listed
  • Ultrasonic Cleaner with Digital Timer
    Ultrasonic Cleaner with Digital Timer
    97043-960
    1.9 L (0.5 gal.)
    117 V, 60 Hz
    L×W×D: 15×14×10 cm (5²⁹⁄₃₂×5½×3¹⁵⁄₁₆")
    Each (1UN)
    Each (1UN) - $6,555.98
    $6,555.98
    Each (1UN)
       $6,555.98
    cUL Listed
    UL Listed
    Accessories
    Specifications
    Environmentally Preferable
    Frequency
    35 kHz
    Temperature range
    20 to 69°C (68 to 156°F)
    Timer
    1 - 99 minutes
    cUL Listed
    UL Listed
  • Ultrasonic Cleaner with Digital Timer
    Ultrasonic Cleaner with Digital Timer
    97043-980
    20.8 L (5.5 gal.)
    117 V, 60 Hz
    L×W×D: 50×30×15 cm (19¹¹⁄₁₆×11¹³⁄₁₆×5²⁹⁄₃₂")
    Each (1UN)
    Each (1UN) - $33,899.28
    $33,899.28
    Each (1UN)
       $33,899.28
    cUL Listed
    UL Listed
    Accessories
    Specifications
    Environmentally Preferable
    Frequency
    35 kHz
    Temperature range
    20 to 69°C (68 to 156°F)
    Timer
    1 - 99 minutes
    cUL Listed
    UL Listed
  • Ultrasonic Cleaner with Digital Timer
    Ultrasonic Cleaner with Digital Timer
    97043-972
    9.5 L (2.5 gal.)
    117 V, 60 Hz
    L×W×D: 30×24×15 cm (11¹³⁄₁₆×9⁷⁄₁₆×5²⁹⁄₃₂")
    Each (1UN)
    Each (1UN) - $22,066.51
    $22,066.51
    Each (1UN)
       $22,066.51
    cUL Listed
    UL Listed
    Accessories
    Specifications
    Environmentally Preferable
    Frequency
    35 kHz
    Temperature range
    20 to 69°C (68 to 156°F)
    Timer
    1 - 99 minutes
    cUL Listed
    UL Listed
Ultrasonic Cleaner with Digital Timer and Heater
  •  
    Capacity
    Electrical
    Tank Dimensions
  • Ultrasonic Cleaner with Digital Timer and Heater
    Ultrasonic Cleaner with Digital Timer and Heater
    97044-010
    20.8 L (5.5 gal.)
    117 V, 60 Hz
    L×W×D: 50×30×15 cm (19¹¹⁄₁₆×11¹³⁄₁₆×5²⁹⁄₃₂")
    Each (1UN)
    Each (1UN) - $39,709.05
    $39,709.05
    Each (1UN)
       $39,709.05
    cUL Listed
    UL Listed
    Accessories
    Specifications
    Environmentally Preferable
    Frequency
    35 kHz
    Temperature range
    20 to 69°C (68 to 156°F)
    Timer
    1 - 99 minutes
    cUL Listed
    UL Listed
  • Ultrasonic Cleaner with Digital Timer and Heater
    Ultrasonic Cleaner with Digital Timer and Heater
    97043-988
    1.9 L (0.5 gal.)
    117 V, 60 Hz
    L×W×D: 15×14×10 cm (5²⁹⁄₃₂×5½×3¹⁵⁄₁₆")
    Each (1UN)
    Each (1UN) - $8,314.91
    $8,314.91
    Each (1UN)
       $8,314.91
    cUL Listed
    UL Listed
    Accessories
    Specifications
    Environmentally Preferable
    Frequency
    35 kHz
    Temperature range
    20 to 69°C (68 to 156°F)
    Timer
    1 - 99 minutes
    cUL Listed
    UL Listed
  • Ultrasonic Cleaner with Digital Timer and Heater
    Ultrasonic Cleaner with Digital Timer and Heater
    97044-002
    9.5 L (2.5 gal.)
    117 V, 60 Hz
    L×W×D: 30×24×15 cm (11¹³⁄₁₆×9⁷⁄₁₆×5²⁹⁄₃₂")
    Each (1UN)
    Each (1UN) - $26,442.48
    $26,442.48
    Paleta de 12 (1 X 1UN)
    Paleta de 12 (1 X 1UN) - $28,822.31
    $28,822.31
    Each (1UN)
       $26,442.48
    Paleta de 12 (1 X 1UN)
       $28,822.31
    cUL Listed
    UL Listed
    Accessories
    Specifications
    Environmentally Preferable
    Frequency
    35 kHz
    Temperature range
    20 to 69°C (68 to 156°F)
    Timer
    1 - 99 minutes
    cUL Listed
    UL Listed
  • Ultrasonic Cleaner with Digital Timer and Heater
    Ultrasonic Cleaner with Digital Timer and Heater
    97043-992
    2.8 L (0.7 gal.)
    117 V, 60 Hz
    L×W×D: 24×14×10 cm (9⁷⁄₁₆×5½×3¹⁵⁄₁₆")
    Each (1UN)
    Each (1UN) - $13,538.37
    $13,538.37
    Each (1UN)
       $13,538.37
    cUL Listed
    UL Listed
    Accessories
    Specifications
    Environmentally Preferable
    Frequency
    35 kHz
    Temperature range
    20 to 69°C (68 to 156°F)
    Timer
    1 - 99 minutes
    cUL Listed
    UL Listed
  • Ultrasonic Cleaner with Digital Timer and Heater
    Ultrasonic Cleaner with Digital Timer and Heater
    97044-014
    28.4 L (7.5 gal.)
    117 V, 60 Hz
    L×W×D: 50×30×20 cm (19¹¹⁄₁₆×11¹³⁄₁₆×7⅞")
    Each (1UN)
    Each (1UN) - $49,424.89
    $49,424.89
    Each (1UN)
       $49,424.89
    cUL Listed
    UL Listed
    Accessories
    Specifications
    Environmentally Preferable
    Frequency
    35 kHz
    Temperature range
    20 to 69°C (68 to 156°F)
    Timer
    1 - 99 minutes
    cUL Listed
    UL Listed
Ultrasonic Cleaner with Digital Timer, Degas, and Heater
  •  
    Capacity
    Electrical
    Tank Dimensions
  • Ultrasonic Cleaner with Digital Timer, Degas, and Heater
    Ultrasonic Cleaner with Digital Timer, Degas, and Heater
    97043-940
    5.7 L (1.5 gal.)
    117 V, 60 Hz
    L×W×D: 30×15×15 cm (11¹³⁄₁₆×5²⁹⁄₃₂×5²⁹⁄₃₂")
    Each (1UN)
    Each (1UN) - $21,906.60
    $21,906.60
    Each (1UN)
       $21,906.60
    cUL Listed
    UL Listed
    Accessories
    Specifications
    Environmentally Preferable
    Frequency
    35 kHz
    Temperature range
    20 to 69°C (68 to 156°F)
    Timer
    1 - 99 minutes
    cUL Listed
    UL Listed
  • Ultrasonic Cleaner with Digital Timer, Degas, and Heater
    Ultrasonic Cleaner with Digital Timer, Degas, and Heater
    97043-948
    13.8 L (3.6 gal.)
    117 V, 60 Hz
    L×W×D: 33×30×15 cm (13×11¹³⁄₁₆×5²⁹⁄₃₂")
    Each (1UN)
    Each (1UN) - $38,429.82
    $38,429.82
    Each (1UN)
       $38,429.82
    cUL Listed
    UL Listed
    Accessories
    Specifications
    Environmentally Preferable
    Frequency
    35 kHz
    Temperature range
    20 to 69°C (68 to 156°F)
    Timer
    1 - 99 minutes
    cUL Listed
    UL Listed
  • Ultrasonic Cleaner with Digital Timer, Degas, and Heater
    Ultrasonic Cleaner with Digital Timer, Degas, and Heater
    97043-944
    9.5 L (2.5 gal.)
    117 V, 60 Hz
    L×W×D: 30×24×15 cm (11¹³⁄₁₆×9⁷⁄₁₆×5²⁹⁄₃₂")
    Each (1UN)
    Each (1UN) - $27,929.59
    $27,929.59
    Each (1UN)
       $27,929.59
    cUL Listed
    UL Listed
    Accessories
    Specifications
    Environmentally Preferable
    Frequency
    35 kHz
    Temperature range
    20 to 69°C (68 to 156°F)
    Timer
    1 - 99 minutes
    cUL Listed
    UL Listed
  • Ultrasonic Cleaner with Digital Timer, Degas, and Heater
    Ultrasonic Cleaner with Digital Timer, Degas, and Heater
    97043-932
    1.9 L (0.5 gal.)
    117 V, 60 Hz
    L×W×D: 15×14×10 cm (5²⁹⁄₃₂×5½×3¹⁵⁄₁₆")
    Each (1UN)
    Each (1UN) - $9,647.44
    $9,647.44
    Each (1UN)
       $9,647.44
    cUL Listed
    UL Listed
    Accessories
    Specifications
    Environmentally Preferable
    Frequency
    35 kHz
    Temperature range
    20 to 69°C (68 to 156°F)
    Timer
    1 - 99 minutes
    cUL Listed
    UL Listed
  • Ultrasonic Cleaner with Digital Timer, Degas, and Heater
    Ultrasonic Cleaner with Digital Timer, Degas, and Heater
    97043-956
    28.4 L (7.5 gal.)
    117 V, 60 Hz
    L×W×D: 50×30×20 cm (19¹¹⁄₁₆×11¹³⁄₁₆×7⅞")
    Each (1UN)
    Each (1UN) - $59,963.32
    $59,963.32
    Each (1UN)
       $59,963.32
    cUL Listed
    UL Listed
    Accessories
    Specifications
    Environmentally Preferable
    Frequency
    35 kHz
    Temperature range
    20 to 69°C (68 to 156°F)
    Timer
    1 - 99 minutes
    cUL Listed
    UL Listed
  • Ultrasonic Cleaner with Digital Timer, Degas, and Heater
    Ultrasonic Cleaner with Digital Timer, Degas, and Heater
    97043-936
    2.8 L (0.7 gal.)
    117 V, 60 Hz
    L×W×D: 24×14×10 cm (9⁷⁄₁₆×5½×3¹⁵⁄₁₆")
    Each (1UN)
    Each (1UN) - $17,100.20
    $17,100.20
    Paleta de 36 (1 X 1UN)
    Paleta de 36 (1 X 1UN) - $18,639.22
    $18,639.22
    Each (1UN)
       $17,100.20
    Paleta de 36 (1 X 1UN)
       $18,639.22
    cUL Listed
    UL Listed
    Accessories
    Specifications
    Environmentally Preferable
    Frequency
    35 kHz
    Temperature range
    20 to 69°C (68 to 156°F)
    Timer
    1 - 99 minutes
    cUL Listed
    UL Listed
  • Ultrasonic Cleaner with Digital Timer, Degas, and Heater
    Ultrasonic Cleaner with Digital Timer, Degas, and Heater
    97043-952
    20.8 L (5.5 gal.)
    117 V, 60 Hz
    L×W×D: 50×30×15 cm (19¹¹⁄₁₆×11¹³⁄₁₆×5²⁹⁄₃₂")
    Each (1UN)
    Each (1UN) - $47,064.57
    $47,064.57
    Each (1UN)
       $47,064.57
    cUL Listed
    UL Listed
    Accessories
    Specifications
    Environmentally Preferable
    Frequency
    35 kHz
    Temperature range
    20 to 69°C (68 to 156°F)
    Timer
    1 - 99 minutes
    cUL Listed
    UL Listed