VWR® CryoPro® Liquid Dewars, L Series
Proveedor: VWR
VWR® CryoPro®
L-4 L-30 L-50 L-20 L-10 L-5
55709-240EA 49304.81 MXN
55709-240 82021-112 55709-242 55709-234 55709-236 55709-238
VWR® CryoPro® Liquid Dewars, L Series
Dewars y recipientes criogénicos
The L series liquid dewars are designed for efficient and safe handling of liquid nitrogen.
  • Convenient storage and dispensation of liquid nitrogen
  • Accessories allow for customization of dewars depending on application
Large or small quantities of liquid nitrogen can be conveniently stored and dispensed with these dewars and accessories. Accessories for pouring and transferring liquid nitrogen allow you to customize the dewars to your application. The phase separator (55709-278) can be attached to the stainless steel transfer hose (55709-244) for increased efficiency and safety when transferring liquid nitrogen.