VWR® Micro Cover Glasses, Round, No. 1
Proveedor: VWR International
VWR®
22CIR-1-601640G 18CIR-1-601640G 18CIR-1-601640G 22CIR-1-601640G 25CIR-1-601640G
48380-046CS 21659.48 MXN
CA48380-068 CA48380-046 48380-046 48380-068 48380-080
VWR® Micro Cover Glasses, Round, No. 1
Cubreobjetos
These cover glasses offer maximum clarity, superior corrosion resistance, and uniform thickness and size.
  • Circular. No. 1 micro cover glasses have a thickness of 0.13 to 0.17mm
They are free of striae and oily film.
Información para pedidos: Packaged 1 oz. per plastic box.