VWR® Kettle Cover Clamps for Resin Reaction Flasks
Proveedor: VWR
VWR®
30392366 30392365
36393-030EA 5977.45 MXN
36393-030 36393-051
VWR® Kettle Cover Clamps for Resin Reaction Flasks
Recipientes de reacción Matraces de Reacción
Provide a convenient, positive, quick-acting means for holding flask and cover together firmly.
  • Convenient for holding flask and cover together
  • Stainless steel springs
Consists of a ring and three insulated, spring-activated clamping arms that fit beneath the flange of the flask body.