Reaction Vessels, Chemglass
Proveedor: Chemglass
CG-1932-02 CG-1920-04 CG-1932-01 CG-1920-01 CG-1932-06 CG-1920-05 CG-1932-05 CG-1932-03 CG-1920-02 CG-1920-06 CG-1932-04 CG-1932-07 CG-1920-03
80071-454EA 10074.57 MXN
80071-538 80071-536 80071-448 80071-456 80071-446 80071-534 80071-454 80071-532 80071-452 80071-540 80071-530 80071-450 80071-528
Reaction Vessels, Chemglass
Recipientes de reacción Vasos de Reacción Cilíndricos
Reaction vessels are constructed with a flange.
The flange is ground and features a groove formed in the sealing surface to allow for a greased seal with an O-ring reaction vessel lid.