VWR® Premium Plain and Frosted Micro Slides
Proveedor: VWR
VWR®
2950-601640T 2956FT-006 631-601640T 2951-601640T 2950FX-601640 2954F-601640 630-601640T 2951DF 2954-001 2954F-601640
48300-026CS 7805.95 MXN
48382-174 75799-268 48300-041 48300-031 48300-026 48312-004 48300-048 75799-274 48300-037 CA48300-036
VWR® Premium Plain and Frosted Micro Slides
Portaobjetos Portaobjetos de microscopios
These pre-cleaned slides are made from the highest-quality sheet glass.
  • Plain and frosted formats available
  • Standard and Economy formats available
  • Glass fabrication
Frosted slides are sandblasted on one end of one side for marking specimens. Double frosted slides are sandblasted on one end of both sides. Economy slides are made from a lesser-quality sheet glass for greater cost-effectiveness.