VWR® Premium VistaVision™ Plain and Frosted Microscope Slides
Proveedor: VWR
VWR® VistaVision™
231150 23110 27525151 23111 231152 23112 231151 2752512 2752510 27525150 2752511VWR
16005-106CS 10905.97 MXN
16004-370 16004-418 16004-426 16004-424 16005-106 16004-422 16005-112 16004-368 16004-420 16004-430 16004-372
VWR® Premium VistaVision™ Plain and Frosted Microscope Slides
Portaobjetos Portaobjetos de microscopios
Microscope slides are manufactured of soda-lime glass in accordance with ISO 8037/1 standards.
  • Manufactured in Germany using high-quality, white soda lime glass
  • Pre-cleaned and ready for use
  • Autoclavable
  • Plain and frosted formats available
A thorough automatic cleaning process ensures that slides are ready to use without further treatment. Frosted slides feature a fine 20 mm frosted marking area.