Micro Slide Boxes, Wood Frame, Jordan Paper Box Company
MODEL B2
48452-001CS 26989.65 MXN
48452-001
Micro Slide Boxes, Wood Frame, Jordan Paper Box Company
Archivadores para portaobjetos
This durable box can hold up to 100 standard slides
It features a clearly numbered plastic insert that provides generous spacing for easy handling of slides. Wood frame is covered with heavy black embossed paper. Hinges and latch are nickel plated. A numbered index lines the cover.