VWR® Economy Microscope Slide Boxes
Proveedor: Avantor
VWR®
HS15991AV HS15990AV HS15991BV HS15991CV HS15990BV HS15990CV
82024-606CS 11782.11 MXN
82024-612 82024-610 82024-616 82024-606 82024-614 82024-608
VWR® Economy Microscope Slide Boxes
Archivadores para portaobjetos
Constructed of durable polypropylene, boxes hold either 25 or 100 standard 25x75 mm (1x3") slides.
  • Color-coded
  • Boxed are unlined
Hinged lids are held securely closed by latch locks. Convenient, space-saving design with stacking feet maximizes stability when boxes are stored atop one another. Color-coded for ease in sorting and selection.