Microbiological Media in Ampoules, Cytiva (Formerly Pall Lab)
Proveedor: Cytiva (Formerly Pall Lab)
68106 4302-N 68108 68100 4302-N 68105 68111 68108 4352 4352 4313-N 68107 4306-N 68101 68102 4307 4307 4306-N 4313-N 68102 68109 68109 68111 68101 68107 68105 68100 68106
28145-686PK 3365.97 MXN
CA28145-680 21432-940 CA28150-502 CA28150-500 CA28150-506 CA28150-504 CA28145-602 CA28145-700 CA28145-686 CA28145-740 28145-695 28145-696 CA28145-684 28145-697 28145-680 CA21432-940 28150-504 28150-502 28150-506 28150-500 CA28145-697 CA28145-696 28145-700 28145-602 28145-684 28145-740 CA28145-695 28145-686
Microbiological Media in Ampoules, Cytiva (Formerly Pall Lab)
Medios Medios preparados
A wide variety of sterile ampoule media for common microbiological analyses, in clear glass or plastic ampoules. Supplied pre-measured for maximum efficiency and convenience. For public wastewater and drinking water: MF-Endo and M-FC (with and without rosolic acid for helping to increase the specificity of the medium for faecal coliforms) media are used when analysing water samples for total coliforms, faecal coliforms and E. coli. For food and beverage: M-TGE, HPC, M-Green YM, and orange serum broths are used in the food and beverage industry to QC final products (and fluids used in the manufacture of these products) to detect potential spoilage organisms. Trypticase soy broth, M-TGE, and HPC may be used to test final products and process waters for total bacteria in the pharmaceutical and microelectronics industries. Pseudomonas and KF-Streptococcal broths are highly selective media for organisms in these genera, and are used to monitor samples wherever contamination by these organisms must be prevented.
  • 2 ml ampoules are a practical, time saving aid for culturing microorganisms on membrane filters
  • Ready to use, no dilutions necessary
  • Shelf life up to 1 year storing at 2 to 8 °C
  • Meet Membrane Filter (MF) Technique Standard Methods requirements with 2 ml ampoules (requirements referenced in Standard Methods for the Examination of Water and Wastewater, 1992 APHA, AWWA, WEF, 20th Edition)