HyClone™ Ham's F-12 Basal Media, Cell Culture Media, Liquid, Cytiva
Proveedor: HyClone products (Cytiva)
HyClone™
SH30026.FS SH30026.02 SH30026.01 SH30526.01 SH30026.02 SH30026.01 SH30026.FS SH30526.01
CA16777-142 16777-142 CA16777-141 16750-096 CA16750-096 16777-141 82003-696 CA82003-696
HyClone™ Ham's F-12 Basal Media, Cell Culture Media, Liquid, Cytiva
Medios de cultivo celular
Ham's F-12 media was originally derived by modifying Ham's F-10 to include increased concentrations of choline, inositol, putrescine, and several amino acids. It was developed specifically for serum free culturing of Chinese Hamster Ovary (CHO) cells and has also been used with supplementation to grow chondrocytes, primary rat hepatocytes, rat prostate epithelial cells, carcinoma cells, rat skeletal myoblasts and rat, rabbit, and chicken embryos.
  • Hydrated using WFI quality water
  • 0,1 μm sterile filtered
  • Manufactured using ISO 9001:2000 certified processes
A Certificate of Analysis is included in each package with preparation instructions, intended use, and handling and performance criteria.
Información para pedidos: Liquid media are supplied in unbreakable, square PETG bottles.