VWR® 80-Place Storage System
2600-227-000 2600-228-000 2600-220-000 2600-223-000 2600-224-000 2600-222-000 2600-226-000 2600-221-000 2600-229-000
30128-282PK 617.82 MXN
30128-270 30128-282 30128-272 30128-274 30128-276 30128-266 30128-278 30128-267 30128-269
VWR® 80-Place Storage System
Storage rack holds 1.7 mL and 2.0 mL microcentrifuge tubes, as well as 2.0 mL cryogenic storage vials.
  • Autoclavable
  • Freezable
Rack and storage box are fully autoclavable and freezable.
Información para pedidos: Racks are available in eight colors and an assorted color pack of five colors: blue, green, orange, red, and yellow. Hinged storage box to store racks is sold separately.