Whatman™ Cellulose Soxhlet Extraction Thimble, Whatman products (Cytiva)
Proveedor: Whatman products (Cytiva)
Whatman™
2810-166 2810-339 2810-166 2800-373 2800-300 2800-250 2810-338 2800-288 2800-280 2800-331 2800-199 2800-280 2800-412 2800-300 2810-266 2800-259 2800-199 2800-226 2800-432 2800-228 2800-308 2800-185 2810-228 2800-330 2800-258 2800-330 2800-608 2800-331 2810-266 2800-185 2800-288 2800-282 2800-412 2800-339 2800-282 2800-432 2800-105 2800-338 2800-266 2810-432 2800-226 2800-260 2800-373 2800-339 2810-902 2810-258 2800-260 2810-258
27732-065EA 162.47 MXN
27732-065 CA11013-710 27732-087 CA27730-142 CA27730-161 27732-145 27732-123 CA27730-140 27732-167 CA28297-430 CA28297-432 27730-117 CA11012-856 CA27730-048 10035-394 76332-602 CA11012-824 70250-016 10035-396 10035-390 10035-392 CA70250-022 10035-398 CA11013-722 27730-048 28297-436 CA11013-720 27730-026 27730-142 28297-432 CA27732-087 CA11012-860 27730-106 27730-128 CA11012-864 CA70250-016 CA11012-858 10035-402 70250-022 27730-060 28297-430 10035-404 27730-140 27730-084 27730-183 27730-161 27730-081 10035-400
Whatman™ Cellulose Soxhlet Extraction Thimble, Whatman products (Cytiva)
Aparatos de extracción Dedales de extracción
Cytiva's Whatman cellulose thimbles are manufactured from high-quality alpha cellulose cotton linter for high-performance use in automated Soxhlet extractor systems
  • Used in a wide variety of solid-liquid extraction studies, including lipid and pesticide analyses in food and soil samples
  • Suitable for temperatures up to 120°C (248°F)
  • Excellent mechanical strength and retention
  • Wall thickness of standard thimbles is 1 to 1.5 mm to support application need
  • Option for double thickness for added rigidity
Standard extraction thimbles have a wall thickness of 1 to 1.5 mm. Double-thickness thimbles have a wall thickness of approximately 2 mm for applications that require higher retention, increased wet or dry strength, or increased rigidity. Measurements can be matched to specific Soxhlet extractor systems.
Used in air and waste gas analysis.