Nalgene® Centrifuge Tubes, Polypropylene Copolymer (PPCO), Conical-Bottom, Graduated, Thermo Scientific
Proveedor: Thermo Scientific
Nalgene®
3103-0050 3103-0015
21008-962CS 17583.72 MXN
21008-973 21008-962
Nalgene® Centrifuge Tubes, Polypropylene Copolymer (PPCO), Conical-Bottom, Graduated, Thermo Scientific
Tubos Tubos de centrífuga
These tubes offer excellent chemical resistance.
  • Autoclavable
  • Maximum RCF: 6000 ×g
  • For proper performance, fill to 80% of total capacity
They incorporate molded graduations. PPCO has a better low temperature rating than standard polypropylene.
Información para pedidos: Supplied without closures. Tube 21008-962 uses closure 21010-894. Tube 21008-973 uses closure 21010-954.