VWR® Magnetized Pipette Rack
Proveedor: Avantor
VWR®
HS20612MV
82007-220EA 1819.86 MXN
82007-220
VWR® Magnetized Pipette Rack
Soportes de pipetas
Durable blue ABS plastic storage rack features circular magnets for secure mounting to metal fume hoods, refrigerators, file cabinets, and more.
  • Durable construction
  • Blue color
  • Magnetized
Four angled compartments accommodate 0.1 mL to 25 mL pipettes. Includes two movable attachments to hold pipetting devices. Attachment dimensions: 10.5Lx3Hcm (41/8x11/4").