VWR® Culture Tubes, Disposable, Borosilicate Glass
Proveedor: VWR
VWR®
20825.430 20825.406 20825.402 20825.425 20825.447 20825.414 20825.443 20825.435
60825-406CS 1223.21 MXN
60825-443 60825-430 60825-406 60825-425 60825-447 60825-414 60825-402 60825-435
VWR® Culture Tubes, Disposable, Borosilicate Glass
Tubos Tubos para cultivo
These tubes are chemically clean and well-suited for general use in chemistry, blood banks, hematology, and other laboratory procedures.
  • Made of borosilicate glass 7.0
  • Fire-polished top
  • Well-rounded bottoms
  • Shrink-film sealed to prevent contamination
Each tube is annealed at temperatures over 426.7 °C (800 °F) to increase strength and reduce breakage.
Tubes are packed in lint-free packaging and are shrink-film sealed to prevent any contamination from outside sources.
Tubes feature smooth, well-rounded bottoms with no irregularities. Tops are fire-polished to eliminate rough edges.