PYREX® Disposable Round Bottom Culture Tubes, White Marking Spot, Screw Cap Style, Bulk Pack, Corning
Proveedor: Corning
PYREX®
99447-20X 99447-161 99447-16X 99447-13 99447-20 99447-16
89090-052 60826-370 60826-381 89090-058 60826-439 89090-056
PYREX® Disposable Round Bottom Culture Tubes, White Marking Spot, Screw Cap Style, Bulk Pack, Corning
Tubos Tubos para cultivo
Designed for both tissue culture and general bacteriological work. Tubes are made of borosilicate glass and are sold without caps.