KIMAX® Test Tubes, Reusable, Borosilicate Glass, with Glass Stopper, Kimble Chase
Proveedor: DWK Life Sciences
KIMAX®
45100-16150 45100-19150 45100-25200 45100-13100
89000-864CS 28093.3 MXN
89000-864 89001-510 89000-866 89001-512
KIMAX® Test Tubes, Reusable, Borosilicate Glass, with Glass Stopper, Kimble Chase
Tubos Tubos para cultivo
Test tubes made from KG-33 borosilicate glass with a [ST] top and a [ST] glass stopper.
Uniform wall thickness for maximum service. Tubes incorporate a durable white ceramic marking spot. Designed from ASTM Specification E982, Type III requirements.