PYREX® Culture Tube, Screw Cap, Corning
Proveedor: Corning
PYREX®
9825-20 9825-20X 9825-16 9825-13 9825-16X 9825-25X 9825-38 9825-25 9825-16XX 9825-22
60827-023CS 3831.1 MXN
60827-045 60827-023 60827-067 60827-089 89091-462 60827-125 60827-147 89090-930 60827-207 60827-205
PYREX® Culture Tube, Screw Cap, Corning
Tubos Tubos para cultivo
Furnished with deep-form phenolic caps to facilitate handling and sealing after autoclaving.
Caps are fabricated from a special formula resistant to the effects of temperature and steam in autoclaving. White enamel marking spot allows excellent surface for pencil notations. Pencil marks are easily erased.
For phenolic caps, see Cat. No. 9999.
Información para pedidos: Choose from tubes with rubber-lined screw caps or fluoropolymer resin-faced rubber-lined screw caps.