PYREX® Separatory Funnels, Squibb, Pear-Shape, [ST] Stopcock, [ST] Stopper, Corning
Proveedor: Corning
PYREX®
6400-2L 6400-500 6400-4L 6400-6L 6400-60 6400-30 6400-1L 6400-125 6400-250
89091-260CS 20300 MXN
89091-512 89091-138 89091-260 30356-008 89091-428 26307-516 30356-100 30356-121 30356-143
PYREX® Separatory Funnels, Squibb, Pear-Shape, [ST] Stopcock, [ST] Stopper, Corning
Embudos Embudos de separación
PYREX®, borosilicate glass, with hollow glass stopper.
With socket and plain stem. With interchangeable glass key.
Stopcock bore, body opening and stem are carefully aligned for even liquid flow with fast cut-off. With smooth tooled tip.