KIMBLE® RAY-SORB® Amber Separatory Funnels, DWK Life Sciences
Proveedor: DWK Life Sciences
KIMBLE® RAY-SORB®
29052F-125 29052F-500 29052F-250
30353-032EA 2310.54 MXN
30353-010 30353-054 30353-032
KIMBLE® RAY-SORB® Amber Separatory Funnels, DWK Life Sciences
Embudos Embudos de separación
Designed with large lower stem I.D
for more complete separation. RAY-SORB® processed to protect materials sensitive to light of shorter wavelengths. Color-coded stopper. Designed from ASTM Specification E1096, Type IV requirements.