SP Wilmad-LabGlass Squibb Separatory Funnels with Glass Stopper and PTFE Plug, Drip Tip, SP Industries
Proveedor: SP Industries
LG-8431T-104 ML-1451T-762 LG-8431T-102 LG-8431T-108
14203-264EA 3954.69 MXN
14206-052 14207-178 14203-486 14203-264
SP Wilmad-LabGlass Squibb Separatory Funnels with Glass Stopper and PTFE Plug, Drip Tip, SP Industries
Embudos Embudos de separación
Featuring bottom stopcock with PTFE plug and glass stopper, these Squibb separatory funnels have a lower standard taper inner joint with a sealed-in drip tube.