Orion™ ISE Filling Solutions, Thermo Scientific
Proveedor: Thermo Scientific
Orion™
900065 900067 900018-WA 900062 900017 900010 951202 900012 900063 900061 951209 900046 951209
34117-967PK 2772.96 MXN
15023-338 34117-967 34117-965 34184-609 34109-157 34117-969 34113-948 CA97008-306 34117-963 34117-961 34108-022 34108-026 97008-306
Orion™ ISE Filling Solutions, Thermo Scientific
Electrolitos Electrolitos de relleno
Enhance the performance and extend the life of ion selective electrodes with Orion™ ISE Filling Solutions.
  • Filling/maintenance solutions are for use with Orion™ Ion Selective Electrodes (ISE)
  • Optimum Result™ ISE filling solutions are specially designed for the Ionplus™ series ion selective electrodes
These solutions are specially formulated to work with the recommended ion selective electrodes and prevent electrode interferences from filling solution.