PYREX® Culture Tubes, Reusable, Borosilicate Glass, Corning
Proveedor: Corning
PYREX®
9820-16X 9820-25X 9820-16XX 9820-25 9820-18 9820-12 9820-22 9820-16 9820-6 9820-10 9820-13 9820-20
60820-320CS 9148.93 MXN
60820-320 60820-265 60820-321 60820-068 60820-104 60820-126 60820-206 60820-228 60820-080 60820-181 60820-261 60820-240
PYREX® Culture Tubes, Reusable, Borosilicate Glass, Corning
Tubos Tubos para cultivo
Made of PYREX® borosilicate glass, rimless.
  • Very good chemical resistance
  • Very good temperature resistance
  • Ends are fire-polished
Culture tubes have walls and bottom of a uniform thickness. All tubes feature a white marking spot, with the exception of 6 and 22 mm tubes.
Tube 5 ml can be used for Kahn tests. Tube 9 ml can be used for Wasserman tests.