PYREX® VISTA™ Rimless Culture Tubes, Corning
Proveedor: Corning
PYREX®
70820-10 70820-16XX 70820-22 70820-25X 70820-18 70820-16X 70820-12 70820-6 70820-25
89090-320CS 4435.57 MXN
89090-414 89090-404 89090-410 89090-400 89090-336 89090-296 89090-364 89090-320 89090-376
PYREX® VISTA™ Rimless Culture Tubes, Corning
Tubos Tubos para cultivo
Borosilicate glass.
Reusable rimless culture tubes offer greater convenience in plugging and rack storage. Round bottom tubes feature fire polished ends. All tubes have a large blue enamel marking spot for simple identification, with the exception of 50, 55 and 70 ml sizes.
Información de suministro: Supplied without caps.