VWR® Microcentrifuge Tubes, PP
Proveedor: VWR
VWR®
3310-E 3315 3300-E 3309 3305 3303-E 3307 3298 3304-E 3299 3297 3316 3312-E 3306 3311-E 3314 3308 3313-E 3302 3296-E 3301
87003-290CS 7323.16 MXN
87003-290 87004-268 87003-292 87004-258 87003-294 87004-266 87004-256 87004-278 87004-264 87003-296 87004-276 87004-254 87003-298 87004-262 87004-274 87004-260 87004-282 87003-300 87004-272 87004-280 87004-270
VWR® Microcentrifuge Tubes, PP
Tubos Tubos de microcentrífugas
Polypropylene tubes feature superior clarity and a positive seal design that withstands repeated cap closures and will not pop open during boiling.
  • Secure positive seal design
  • Tubes fit all standard rotors
  • Certified RNase-, DNase-, and pyrogen-free
  • Certified PCR inhibitor- and human DNA-free
  • Autoclavable
Temperature range is from –80 to 121 °C (–112 to 250 °F).
Información para pedidos: Rainbow packs consist of red, blue, green, yellow, and purple tubes.