VWR® PCR Tubes and Caps
Proveedor: VWR
VWR®
3220-00-111AV 3320-00-111AV 3210-09-111AV 3320-09-111AV 3310-00-111AV 3225-00-111AV 3210-00-111AV
20170-010CS 11581.88 MXN
76446-876 76446-878 76318-804 76446-868 20170-010 20170-012 76446-880
VWR® PCR Tubes and Caps
Tubos Tubos para PCR
Ideal for all PCR protocols, these 0.2 ml tubes and caps are made from polypropylene and are compatible with most popular thermal cycler blocks.
  • Autoclavable
  • Chemically inert
They feature smooth, thin, uniform walls for accurate thermal transfer. Certified free of RNase, DNase, and human DNA (lot-by-lot certificates available). Tubes are available with bubble-top or flat-top caps, both of which fit precisely to ensure a positive, contamination-free environment.