Order Entry
Refracto Portable Refractometers, Mettler-Toledo
  11276-898
Product available on GSA Advantage®
undefined
Refracto Portable Refractometers, Mettler-Toledo
  11276-898
 :  51324650
Product available on GSA Advantage®
$147,108.00
  Each (1UN)
  • Accuracy:
    ±0.0005 nD, ±0.2% Brix
  • Measurement range:
    1.32–1.50 nD (Model 30PX), 1.32–1.65 nD (Model 30GS), 0–85% Brix
  • Resolution:
    0.0001 nD, 0.1% Brix
  • Temperature range:
    10 to 40°C (50 to 104°F)
  • Description:
    Refracto 30PX
  • Operating temperature:
    5...35 °C (41...95 °F)
  • Weight:
    250 g (8.8 oz.)
  • Cat. no.:
    11276-898
  • Supplier No.:
    51324650