Corning® Culture Dishes, Nontreated, Sterile, Corning
Proveedor: Corning
Corning®
430597 430591 431111 430589 431301 430588 431272
77776-742CS 4300 MXN
25382-452 25382-450 25382-470 77776-922 29186-491 77776-742 25382-456
Corning® Culture Dishes, Nontreated, Sterile, Corning
Placas de cultivo Placas para cultivo celular
Optically clear PS. For applications where cell attachment is not desired.
  • With stacking beads to aid handling
  • Supplied with vents to provide consistent gas exchange
  • Sterilized by gamma radiation
  • Certified non-pyrogenic