Petri Dishes, Sterile, Cytiva (Formerly Pall Lab)
Proveedor: Cytiva
7242 7232 7242 7245 7245 7232
25388-640CS 2041.59 MXN
25388-581 CA25388-640 CA25388-606 25388-640 CA25388-581 25388-606
Petri Dishes, Sterile, Cytiva (Formerly Pall Lab)
Placas de cultivo Placas de Petri
PS dish, cellulose pad. Ideal for microbiological analysis when performing the membrane filter technique. Petri dishes with absorbent pads can be used with broth media, or users can pour agar into dishes without absorbent pads. Accepts 47 mm membrane filter.
  • Gamma irradiated so no Ethylene Oxide residuals to impede microbial growth
  • Easy to use, opens easily with one hand, yet closes to a tight seal
  • Uses less space on the laboratory bench or in an incubator with easy stacking base