Petri Dishes and Contact Dishes, Polystyrene, Greiner bio-one
Proveedor: Greiner bio-one
628102 639102 629180 633102 639161 632102 688102 635102 632161 633181 629161 633161 632181 633180 664161 632180 627161 664102 628161 688161 627102
82051-068CS 10334.76 MXN
82051-068 82051-066 82050-914 82050-918 82050-544 82050-566 82050-600 82050-578 82050-536 82050-548 82050-592 82050-570 82050-560 82050-572 82050-562 82050-584 82050-540 82050-596 82050-574 82050-564 82050-586
Petri Dishes and Contact Dishes, Polystyrene, Greiner bio-one
Placas de cultivo Placas de Petri
These polystyrene petri dishes and contact dishes are available in different sizes and formats.
  • Available with or without vents
  • Nonsterile and sterile
  • Highly transparent polystyrene construction
Manufactured from highly transparent polystyrene, dishes are heat-resistant up to 60°C for use with hot agar.
Depending on the application, vented or non-vented petri dishes and contact dishes can be selected. If applicable, sterile dishes are also available.
Besides the standard round design, a square-profile petri dish is also available. Special models include petri dishes with two or three compartments and germ count dishes.