SP Bel-Art Poxygrid® Petri Dish Carrying Racks, 100 mm, Bel-Art Products, a part of SP
Proveedor: Bel-Art
SCIENCEWARE®
H18979-0000 H18978-0000 H18978-0000
62866-192EA 3922.29 MXN
62866-192 62866-206 490012-238
SP Bel-Art Poxygrid® Petri Dish Carrying Racks, 100 mm, Bel-Art Products, a part of SP
Gradillas Dispensadores de placas Petri
Poxygrid® petri dish carrying racks hold 100 x 20 mm plastic petri dishes.
  • Epoxy-coated steel wire is chemical- and corrosion-resistant
  • The rack is steam autoclavable at 121 °C (250 °F)
  • Not suitable for glass dishes
Wide front opening prevents dishes from sliding out during transport but allows easy loading and emptying from the top.