VWR® Petri Dish Racks, 100 mm
Proveedor: VWR
VWR® Easyplate™
18982-6309 18982-6314 18982-6109 18982-6209 18982-6014 18982-6009 18982-6409
10600-108EA 5605.45 MXN
10600-106 10600-108 10600-112 10600-102 10600-114 10600-104 10600-110
VWR® Petri Dish Racks, 100 mm
Gradillas Dispensadores de placas Petri
Increase workspace and efficiently transport large quantity of Petri dishes.
  • Silicone holding strips grip each Petri dish; easily insert and remove groups of dishes without disturbing upper or lower dishes
  • Side labels provide space for critical sample identification
  • Lightweight, chemical resistant polycarbonate
  • Autoclavable at 121 °C (250 °F)
Available in 4 colors and two sizes. The 24.1 cm (9.5") high size holds forty-eight 100 mm Petri dishes, and the 36.8 cm (14.5") high size holds eighty-four 100 mm Petri dishes.
Información para pedidos: Assorted colors pack includes one rack in each of the following colors: white, red, yellow, and blue.