Cytoseal™ Mounting Medium, Epredia™
Proveedor: Epredia
Cytoseal™
8310-4 8311-4 8311-4 8310-16 8310-4
48212-187CS 13403.87 MXN
48212-187 48212-154 CA48212-267 48212-267 CA48212-154
Cytoseal™ Mounting Medium, Epredia™
Tintes Reactivos de microscopía y tinción
Prevent cracking, fading and discoloration of stained specimens with Epredia™ 60 and 280 mounting media.
  • All paraffins melt between 55° and 57 °C and are triple filtered to 0.5 µm
  • Flake form has a greater surface-to-weight ratio than pellets
  • Faster melting time
  • Type 1 contains no polymer and offers rapid infiltration
  • Type 3 contains minimal polymer and offers fast infiltration
  • Type 6 is harder than Type 3 and allows for thinner sections
  • Type 9 has the highest polymer content available and yields extremely thin sections
Clear and colorless, medium is formulated from high-quality advanced acrylic resins. It dries rapidly and does not become brittle and crack or discolor with age. Preserves original slide specimen quality almost indefinitely. Slides mounted with this medium will not stick together, and it can be dissolved in toluene or xylene if tissue requires remounting. Available in two viscosities: Cytoseal™ 60 for use with cover glass, and Cytoseal™ 280 for thicker coating or minimal spreading. Ideal for use with oil immersion objectives and in fluorescent applications.