ThermoSafe® Dry Ice Machine, Sonoco ThermoSafe
560
87003-904EA 34093 MXN
87003-904
ThermoSafe® Dry Ice Machine, Sonoco ThermoSafe
Dry ice maker is ideal for tissue preservation, specimen shipping, biologicals manufacturing, industrial processes, metal fabrication, and classroom demonstrations.
  • Ideal for tissue preservation
  • Simple operation
  • No additional equipment required
Simple operation creates ready-to-use, 1kg ( 2.2lb.) dry ice blocks measuring 15.2Lx10.2Wx3.8Hcm (6x4x11/2") in two minutes. Requires no additional equipment or procedures except the CO2 supply (not included). Internal heat exchange device delivers an efficient use of CO2.
Información para pedidos: Includes one-year manufacturer’s warranty.