KIMAX® Nessler Color Comparison Tubes, Tall Form, APHA Standard, Kimble Chase
Proveedor: DWK Life Sciences
KIMAX®
45315-50100 45315-100 45315A-50 45315-50 45325A-50 45315B-100 45325B-50100 45315A-50100 45315B-50 45315A-100
66176-348CS 2923.24 MXN
66176-348 66176-326 66176-305 16143-254 16143-253 66176-529 66176-507 72640-048 66176-620 66176-686
KIMAX® Nessler Color Comparison Tubes, Tall Form, APHA Standard, Kimble Chase
Tubos Tubos Nessler
These narrow, tall tubes with a long scale length feature 'Shadowless Bottoms' that transmit undistorted light from a source beneath tube. All graduation lines and legends are printed in white ceramic enamel.
  • Ideal for color comparison
  • Shadowless bottom of each tube transmits undistorted light
  • Rings and legend are durable white ceramic enamel
Available unmatched or in matched sets of 6 or 12.
The 50 to 100 ml tubes with cap stopper were designed from ASTM D1209. [ST] 24/12 taper cap style stoppers need not be removed when making readings. The word 'Shadowless' is printed on the side of the stopper to distinguish between these stoppers and regular weighing bottle stoppers of the same size. Tubes come in a matched set of 12, with the exception of 16143-256, which is a matched set of 6.
Graduated ring scales on the 50 ml matched sets do not vary by more than 1.5 mm; graduated scales on the 100 ml matched sets do not vary by more than 2 mm. The 50 ml size is specifically mentioned by American Public Health Association (APHA) in Standard Methods for the Examination of Water and Wastewater. Designed from Federal Specification NNN-T-175, Type 1, Class 2 requirements.